Tisk…

4-Fluoro-3-nitrobenzyl alcohol

Dodavatel: Apollo Scientific

PC502598-25G PC502598-5G
APOSPC502598-25GEA 7280 CZK
APOSPC502598-25G APOSPC502598-5G
4-Fluoro-3-nitrobenzyl alcohol
4-Fluoro-3-nitrobenzyl alcohol
Vzorec: C₇H₆FNO₃
MW: 171,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861396
CAS číslo: 20274-69-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR