Tisk…

2,3-Difluoro-4-(trifluoromethoxy)benzonitrile 95%

Dodavatel: Apollo Scientific
Varování

PC502526-1G PC502526-250MG PC502526-5G
APOSPC502526-1GEA 1660 CZK
APOSPC502526-1G APOSPC502526-250MG APOSPC502526-5G
2,3-Difluoro-4-(trifluoromethoxy)benzonitrile 95%
2,3-Difluoro-4-(trifluoromethoxy)benzonitrile
Vzorec: C₈H₂F₅NO
MW: 223,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22580791
CAS číslo: 1404194-78-0
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR