Tisk…

2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC502521-1G PC502521-250MG
APOSPC502521-1GEA 16140 CZK
APOSPC502521-1G APOSPC502521-250MG
2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde 95%
2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde
Vzorec: C₈H₃F₅O₂
MW: 226,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561154
CAS číslo: 2244086-99-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR