2,3-Difluoro-4-(trifluoromethoxy)benzyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC502518-1G
PC502518-250MG
PC502518-5G
APOSPC502518-1GEA
1660
CZK
APOSPC502518-1G
APOSPC502518-250MG
APOSPC502518-5G
2,3-Difluoro-4-(trifluoromethoxy)benzyl bromide 95%
2,3-Difluoro-4-(trifluoromethoxy)benzyl bromide
Vzorec:
C₈H₄BrF₅O MW: 291,013 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31561153 CAS číslo: 2244078-00-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...