Tisk…

2,6-Difluoro-4-(trifluoromethoxy)benzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC502513-1G PC502513-250MG PC502513-5G
APOSPC502513-1GEA 992 CZK
APOSPC502513-1G APOSPC502513-250MG APOSPC502513-5G
2,6-Difluoro-4-(trifluoromethoxy)benzaldehyde 95%
2,6-Difluoro-4-(trifluoromethoxy)benzaldehyde
Vzorec: C₈H₃F₅O₂
MW: 226,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28399110
CAS číslo: 1799439-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR