3,6-Difluoro-2-(trifluoromethoxy)benzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC502486-1G
PC502486-250MG
PC502486-5G
APOSPC502486-1GEA
1990
CZK
APOSPC502486-1G
APOSPC502486-250MG
APOSPC502486-5G
3,6-Difluoro-2-(trifluoromethoxy)benzonitrile 95%
3,6-Difluoro-2-(trifluoromethoxy)benzonitrile
Vzorec:
C₈H₂F₅NO MW: 223,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28791014 CAS číslo: 1803789-95-8 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...