Tisk…

3,6-Difluoro-2-(trifluoromethoxy)benzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502480-1G PC502480-250MG PC502480-5G
APOSPC502480-1GEA 1660 CZK
APOSPC502480-1G APOSPC502480-250MG APOSPC502480-5G
3,6-Difluoro-2-(trifluoromethoxy)benzyl bromide 95%
3,6-Difluoro-2-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₄BrF₅O
MW: 291,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561161
CAS číslo: 2244076-54-6
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR