Tisk…

1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene

Dodavatel: Apollo Scientific
Varování

PC502465-1G PC502465-5G PC502465-10G
APOSPC502465-1GEA 6220 CZK
APOSPC502465-1G APOSPC502465-5G APOSPC502465-10G
1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene
1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene
Vzorec: C₇H₃BrF₄O
MW: 258,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22551456
CAS číslo: 1129541-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR