3-Bromo-2,5,6-trifluorobenzyl alcohol 98%
Dodavatel: Apollo Scientific
PC502418-1G
PC502418-5G
APOSPC502418-1GEA
6580
CZK
APOSPC502418-1G
APOSPC502418-5G
3-Bromo-2,5,6-trifluorobenzyl alcohol 98%
3-Bromo-2,5,6-trifluorobenzyl alcohol
Vzorec:
C₇H₄BrF₃O MW: 241.005 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30830100 CAS číslo: 1296310-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...