Tisk…

3-Bromo-2,5,6-trifluorobenzyl bromide 97%

Dodavatel: Apollo Scientific

PC502348-1G PC502348-5G
APOSPC502348-1GEA 7940 CZK
APOSPC502348-1G APOSPC502348-5G
3-Bromo-2,5,6-trifluorobenzyl bromide 97%
3-Bromo-2,5,6-trifluorobenzyl bromide
Vzorec: C₇H₃Br₂F₃
MW: 303,902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30830102
CAS číslo: 2149598-68-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR