Tisk…

3-Bromo-2,5,6-trifluorobenzonitrile

Dodavatel: Apollo Scientific

PC502347-1G PC502347-5G
APOSPC502347-1GEA 9260 CZK
APOSPC502347-1G APOSPC502347-5G
3-Bromo-2,5,6-trifluorobenzonitrile
3-Bromo-2,5,6-trifluorobenzonitrile
Vzorec: C₇HBrF₃N
MW: 235,989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30830098
CAS číslo: 485318-78-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR