Tisk…

3-Bromo-4-fluoro-2-nitroanisole 95%

Dodavatel: Apollo Scientific
Varování

PC502342-1G PC502342-250MG PC502342-5G
APOSPC502342-1GEA 1410 CZK
APOSPC502342-1G APOSPC502342-250MG APOSPC502342-5G
3-Bromo-4-fluoro-2-nitroanisole 95%
3-Bromo-4-fluoro-2-nitroanisole
Vzorec: C₇H₅BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD28739837
CAS číslo: 1807192-67-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR