Tisk…

3-Bromo-2-(difluoromethoxy)-5-fluoronitrobenzene

Dodavatel: Apollo Scientific
Varování

PC502337-1G PC502337-250MG
APOSPC502337-1GEA 17360 CZK
APOSPC502337-1G APOSPC502337-250MG
3-Bromo-2-(difluoromethoxy)-5-fluoronitrobenzene
3-Bromo-2-(difluoromethoxy)-5-fluoronitrobenzene
Vzorec: C₇H₃BrF₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD28737947
CAS číslo: 1805591-69-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR