Tisk…

1-(Difluoromethoxy)-4-nitroisoquinoline

Dodavatel: Apollo Scientific
Varování

PC502324-1G PC502324-250MG
APOSPC502324-1GEA 16340 CZK
APOSPC502324-1G APOSPC502324-250MG
1-(Difluoromethoxy)-4-nitroisoquinoline
1-(Difluoromethoxy)-4-nitroisoquinoline
Vzorec: C₁₀H₆F₂N₂O₃
MW: 240.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30730349
CAS číslo: 2187434-99-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR