Tisk…

2-Fluoro-4-iodophenylacetic acid

Dodavatel: Apollo Scientific
Varování

PC50230-1G PC50230-250MG PC50230-5G
APOSPC50230-1GEA 2820 CZK
APOSPC50230-1G APOSPC50230-250MG APOSPC50230-5G
2-Fluoro-4-iodophenylacetic acid
2-Fluoro-4-iodophenylacetic acid
Vzorec: C₈H₆FIO₂
MW: 280,035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396991
CAS číslo: 345963-96-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR