Tisk…

2-(4-Amino-2-fluorophenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

PC50229-1G PC50229-250MG PC50229-5G
APOSPC50229-1GEA 2400 CZK
APOSPC50229-1G APOSPC50229-250MG APOSPC50229-5G
2-(4-Amino-2-fluorophenyl)acetic acid
2-(4-Amino-2-fluorophenyl)acetic acid
Vzorec: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09032899
CAS číslo: 914224-31-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR