Tisk…

2-Fluoro-4-iodophenylacetonitrile

Dodavatel: Apollo Scientific
Varování

PC50227-1G PC50227-250MG PC50227-5G
APOSPC50227-1GEA 2440 CZK
APOSPC50227-1G APOSPC50227-250MG APOSPC50227-5G
2-Fluoro-4-iodophenylacetonitrile
2-Fluoro-4-iodophenylacetonitrile
Vzorec: C₈H₅FIN
MW: 261,035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396999
CAS číslo: 345963-95-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR