3-Ethynyl-2-fluorobenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC502253-1G
PC502253-250MG
PC502253-5G
APOSPC502253-1GEA
12200
CZK
APOSPC502253-1G
APOSPC502253-250MG
APOSPC502253-5G
3-Ethynyl-2-fluorobenzaldehyde 98%
3-Ethynyl-2-fluorobenzaldehyde
Vzorec:
C₉H₅FO MW: 148.134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30730328 CAS číslo: 1851952-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...