5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Dodavatel: Apollo Scientific
|
Varování
|
PC50223-100MG
PC50223-1G
PC50223-250MG
APOSPC50223-100MGEA
3810
CZK
APOSPC50223-100MG
APOSPC50223-1G
APOSPC50223-250MG
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Vzorec:
C₁₀H₁₀F₃N·ClH MW: 237,649 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08544284 CAS číslo: 215788-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...