Tisk…

2-Hydroxy-3-nitro-5-(trifluoromethoxy)benzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC502223-1G PC502223-250MG
APOSPC502223-1GEA 16140 CZK
APOSPC502223-1G APOSPC502223-250MG
2-Hydroxy-3-nitro-5-(trifluoromethoxy)benzaldehyde 95%
2-Hydroxy-3-nitro-5-(trifluoromethoxy)benzaldehyde
Vzorec: C₈H₄F₃NO₅
MW: 251,116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30730319
CAS číslo: 2149597-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR