Tisk…

Potassium (2,4-difluorobenzyl)trifluoroborate

Dodavatel: Apollo Scientific
Varování

PC502211-1G PC502211-250MG
APOSPC502211-1GEA 15510 CZK
APOSPC502211-1G APOSPC502211-250MG
Potassium (2,4-difluorobenzyl)trifluoroborate
Potassium (2,4-difluorobenzyl)trifluoroborate
Vzorec: C₇H₅BF₅·K
Storage Temperature: Ambient
MDL Number: MFCD18861751
CAS číslo: 1632070-90-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR