Tisk…

2,4-Difluoro-3-(trifluoromethoxy)phenacyl bromide 95+%

Dodavatel: Apollo Scientific
Nebezpečí

PC502207-1G PC502207-250MG
APOSPC502207-1GEA 37360 CZK
APOSPC502207-1G APOSPC502207-250MG
2,4-Difluoro-3-(trifluoromethoxy)phenacyl bromide 95+%
2,4-Difluoro-3-(trifluoromethoxy)phenacyl bromide
Vzorec: C₉H₄BrF₅O₂
MW: 319,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723449
CAS číslo: 2149602-08-2
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR