Tisk…

2',4'-Difluoro-3’-(trifluoromethoxy)acetophenone 97%

Dodavatel: Apollo Scientific
Varování

PC502206-1G PC502206-250MG
APOSPC502206-1GEA 17360 CZK
APOSPC502206-1G APOSPC502206-250MG
2',4'-Difluoro-3’-(trifluoromethoxy)acetophenone 97%
2',4'-Difluoro-3’-(trifluoromethoxy)acetophenone
Vzorec: C₉H₅F₅O₂
MW: 240,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723445
CAS číslo: 2149591-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR