5-Bromo-2-(difluoromethoxy)-3-(trifluoromethyl)pyridine 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC502205-1G
PC502205-250MG
PC502205-5G
APOSPC502205-1GEA
14880
CZK
APOSPC502205-1G
APOSPC502205-250MG
APOSPC502205-5G
5-Bromo-2-(difluoromethoxy)-3-(trifluoromethyl)pyridine 95%
5-Bromo-2-(difluoromethoxy)-3-(trifluoromethyl)pyridine
Vzorec:
C₇H₃BrF₅NO Storage Temperature: Ambient |
MDL Number:
MFCD28738216 CAS číslo: 1804896-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...