2,4-Difluoro-3-(trifluoromethoxy)bromobenzene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC502201-1G
PC502201-250MG
APOSPC502201-1GEA
20470
CZK
APOSPC502201-1G
APOSPC502201-250MG
2,4-Difluoro-3-(trifluoromethoxy)bromobenzene 95%
2,4-Difluoro-3-(trifluoromethoxy)bromobenzene
Vzorec:
C₇H₂BrF₅O MW: 276.986 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28738353 CAS číslo: 1807187-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...