Tisk…

Potassium (3-fluorobenzyl)trifluoroborate

Dodavatel: Apollo Scientific
Varování

PC502192-1G PC502192-250MG
APOSPC502192-1GEA 4300 CZK
APOSPC502192-1G APOSPC502192-250MG
Potassium (3-fluorobenzyl)trifluoroborate
Potassium (3-fluorobenzyl)trifluoroborate
Vzorec: C₇H₆BF₄·K
MW: 216,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112714
CAS číslo: 2126821-95-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR