4-Fluoro-3-[(trifluoromethyl)sulphonyl]benzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Fluoro-3-((trifluoromethyl)sulphonyl)benzene-1-sulphonyl chloride
PC502180-1G
PC502180-250MG
PC502180-5G
APOSPC502180-1GEA
2280
CZK
APOSPC502180-1G
APOSPC502180-250MG
APOSPC502180-5G
4-Fluoro-3-[(trifluoromethyl)sulphonyl]benzenesulphonyl chloride
4-Fluoro-3-[(trifluoromethyl)sulphonyl]benzenesulphonyl chloride
Vzorec:
C₇H₃ClF₄O₄S₂ MW: 326.673 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16883063 CAS číslo: 1027345-07-8 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...