3,5-Difluoro-2-nitrobenzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC502155-1G
PC502155-250MG
APOSPC502155-1GEA
18620
CZK
APOSPC502155-1G
APOSPC502155-250MG
3,5-Difluoro-2-nitrobenzyl alcohol 95%
3,5-Difluoro-2-nitrobenzyl alcohol
Vzorec:
C₇H₅F₂NO₃ MW: 189,116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17169029 CAS číslo: 1378491-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...