3-Iodo-5-(trifluoromethyl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC502150-1G
PC502150-250MG
APOSPC502150-1GEA
20830
CZK
APOSPC502150-1G
APOSPC502150-250MG
3-Iodo-5-(trifluoromethyl)benzaldehyde
3-Iodo-5-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₄F₃IO Storage Temperature: Ambient |
MDL Number:
MFCD11977391 CAS číslo: 868166-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...