Perfluorobutylsulphonamide 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,1,2,2,3,3,4,4,4-Nonafluorobutane-1-sulphonamide
PC50215-1G
PC50215-250MG
PC50215-5G
APOSPC50215-1GEA
8270
CZK
APOSPC50215-1G
APOSPC50215-250MG
APOSPC50215-5G
Perfluorobutylsulphonamide 97%
Perfluorobutylsulphonamide
Vzorec:
C₄H₂F₉NO₂S MW: 299,115 g/mol Bod tání: 64…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD11112029 CAS číslo: 30334-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...