Tisk…

2-Bromo-6-(trifluoromethyl)benzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

PC502142-1G PC502142-250MG PC502142-5G
APOSPC502142-1GEA 5260 CZK
APOSPC502142-1G APOSPC502142-250MG APOSPC502142-5G
2-Bromo-6-(trifluoromethyl)benzaldehyde 95%
2-Bromo-6-(trifluoromethyl)benzaldehyde
Vzorec: C₈H₄BrF₃O
MW: 253,016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22381476
CAS číslo: 1236141-95-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR