Tisk…

3,5-Difluoro-2-nitrobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC502141-1G PC502141-250MG
APOSPC502141-1GEA 18620 CZK
APOSPC502141-1G APOSPC502141-250MG
3,5-Difluoro-2-nitrobenzyl bromide 95%
3,5-Difluoro-2-nitrobenzyl bromide
Vzorec: C₇H₄BrF₂NO₂
MW: 252.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17169030
CAS číslo: 1258546-68-7
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR