Tisk…

5-Fluoro-2-(hydroxymethyl)benzeneboronic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Fluoro-2-hydroxymethylphenylboronic acid, (5-Fluoro-2-(hydroxymethyl)phenyl)boronic acid

PC502140-1G PC502140-5G PC502140-10G PC502140-25G
APOSPC502140-1GEA 2650 CZK
APOSPC502140-1G APOSPC502140-5G APOSPC502140-10G APOSPC502140-25G
5-Fluoro-2-(hydroxymethyl)benzeneboronic acid 95%
5-Fluoro-2-(hydroxymethyl)benzeneboronic acid
Vzorec: C₇H₈BFO₃
MW: 169.946 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11111910
CAS číslo: 1246633-53-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR