Tisk…

3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid 97%

Dodavatel: Apollo Scientific

Synonyms: (3-Fluoro-2-(hydroxymethyl)phenyl)boronic acid, 3-Fluoro-2-(hydroxymethyl)phenylboronic acid

PC502136-1G PC502136-5G
APOSPC502136-1GEA 2480 CZK
APOSPC502136-1G APOSPC502136-5G
3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid 97%
3-Fluoro-2-(hydroxymethyl)-benzeneboronic acid
Vzorec: C₇H₈BFO₃
MW: 169,946 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181611
CAS číslo: 1246633-55-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR