Tisk…

Methyl 3-iodo-5-(trifluoromethyl)benzoate 95%

Dodavatel: Apollo Scientific
Varování

PC502131-1G PC502131-250MG PC502131-5G
APOSPC502131-1GEA 1660 CZK
APOSPC502131-1G APOSPC502131-250MG APOSPC502131-5G
Methyl 3-iodo-5-(trifluoromethyl)benzoate 95%
Methyl 3-iodo-5-(trifluoromethyl)benzoate
Vzorec: C₉H₆F₃IO₂
MW: 330.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11976042
CAS číslo: 161622-17-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR