Tisk…

3-Iodo-4-(trifluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

PC502103-1G PC502103-250MG
APOSPC502103-1GEA 10350 CZK
APOSPC502103-1G APOSPC502103-250MG
3-Iodo-4-(trifluoromethyl)benzonitrile
3-Iodo-4-(trifluoromethyl)benzonitrile
Vzorec: C₈H₃F₃IN
MW: 297,016 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18398161
CAS číslo: 1261482-46-5
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR