6-Bromo-5-fluoro-4-(trifluoromethyl)-1H-benzimidazole
Dodavatel: Apollo Scientific
|
Varování
|
PC50208-1G
PC50208-250MG
APOSPC50208-1GEA
13230
CZK
APOSPC50208-1G
APOSPC50208-250MG
6-Bromo-5-fluoro-4-(trifluoromethyl)-1H-benzimidazole
6-Bromo-5-fluoro-4-(trifluoromethyl)-1H-benzimidazole
Vzorec:
C₈H₃BrF₄N₂ MW: 283,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28515425 CAS číslo: 1820619-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...