Tisk…

4-Bromo-2-(trifluoromethoxy)phenylacetylene

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-1-ethynyl-2-(trifluoromethoxy)benzene

PC502072-1G PC502072-250MG
APOSPC502072-1GEA 3410 CZK
APOSPC502072-1G APOSPC502072-250MG
4-Bromo-2-(trifluoromethoxy)phenylacetylene
4-Bromo-2-(trifluoromethoxy)phenylacetylene
Vzorec: C₉H₄BrF₃O
MW: 265,027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22570386
CAS číslo: 1824630-55-8
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR