Methyl 2,6-difluoro-3-(trifluoromethyl)benzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC502071-1G
PC502071-250MG
PC502071-5G
APOSPC502071-1GEA
9530
CZK
APOSPC502071-1G
APOSPC502071-250MG
APOSPC502071-5G
Methyl 2,6-difluoro-3-(trifluoromethyl)benzoate
Methyl 2,6-difluoro-3-(trifluoromethyl)benzoate
Vzorec:
C₉H₅F₅O₂ MW: 240,127 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30481112 CAS číslo: 2089258-73-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...