3-Bromo-2-(trifluoromethyl)benzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC502063-1G
PC502063-250MG
PC502063-5G
APOSPC502063-1GEA
6850
CZK
APOSPC502063-1G
APOSPC502063-250MG
APOSPC502063-5G
3-Bromo-2-(trifluoromethyl)benzonitrile 95%
3-Bromo-2-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₃BrF₃N MW: 250.015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16606444 CAS číslo: 1228898-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...