Tisk…

5-Bromo-2,3-difluorobenzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2,3-difluorobenzene methanol

PC502062-1G PC502062-250MG PC502062-5G
APOSPC502062-1GEA 10350 CZK
APOSPC502062-1G APOSPC502062-250MG APOSPC502062-5G
5-Bromo-2,3-difluorobenzyl alcohol 95%
5-Bromo-2,3-difluorobenzyl alcohol
Vzorec: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972473
CAS číslo: 887585-71-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR