3-Bromo-4,5-difluorobenzonitrile 99+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC502060-1G
PC502060-5G
APOSPC502060-1GEA
1620
CZK
APOSPC502060-1G
APOSPC502060-5G
3-Bromo-4,5-difluorobenzonitrile 99+%
3-Bromo-4,5-difluorobenzonitrile
Vzorec:
C₇H₂BrF₂N Storage Temperature: Ambient |
MDL Number:
MFCD20233294 CAS číslo: 1349715-72-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...