Tisk…

3’-Iodo-5’-(trifluoromethyl)acetophenone

Dodavatel: Apollo Scientific
Varování

PC502023-1G PC502023-250MG PC502023-5G
APOSPC502023-1GEA 20670 CZK
APOSPC502023-1G APOSPC502023-250MG APOSPC502023-5G
3’-Iodo-5’-(trifluoromethyl)acetophenone
3’-Iodo-5’-(trifluoromethyl)acetophenone
Vzorec: C₉H₆F₃IO
MW: 314,043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544410
CAS číslo: 1393557-53-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR