Tisk…

2-Bromo-3,4-difluorobenzaldehyde

Dodavatel: Apollo Scientific
Varování

PC501996-1G PC501996-250MG
APOSPC501996-1GEA 2900 CZK
APOSPC501996-1G APOSPC501996-250MG
2-Bromo-3,4-difluorobenzaldehyde
2-Bromo-3,4-difluorobenzaldehyde
Vzorec: C₇H₃BrF₂O
MW: 220,999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28669844
CAS číslo: 1578156-21-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR