Tisk…

2-Bromo-5-(difluoromethoxy)nitrobenzene 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-4-(difluoromethoxy)-2-nitrobenzene

PC501994-1G PC501994-250MG PC501994-5G
APOSPC501994-1GEA 4960 CZK
APOSPC501994-1G APOSPC501994-250MG APOSPC501994-5G
2-Bromo-5-(difluoromethoxy)nitrobenzene 95%
2-Bromo-5-(difluoromethoxy)nitrobenzene
Vzorec: C₇H₄BrF₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD18390950
CAS číslo: 865886-84-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR