2-Amino-4-bromo-5-fluorobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC501993-1G
PC501993-250MG
APOSPC501993-1GEA
16340
CZK
APOSPC501993-1G
APOSPC501993-250MG
2-Amino-4-bromo-5-fluorobenzonitrile
2-Amino-4-bromo-5-fluorobenzonitrile
Vzorec:
C₇H₄BrFN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD29049266 CAS číslo: 1935427-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...