2-(Difluoromethoxy)-6-fluoronitrobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC501987-1G
PC501987-250MG
PC501987-5G
APOSPC501987-1GEA
1660
CZK
APOSPC501987-1G
APOSPC501987-250MG
APOSPC501987-5G
2-(Difluoromethoxy)-6-fluoronitrobenzene
2-(Difluoromethoxy)-6-fluoronitrobenzene
Vzorec:
C₇H₄F₃NO₃ MW: 207,107 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14698448 CAS číslo: 1214336-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...