2-(Difluoromethoxy)-4-fluoroaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC501980-1G
PC501980-5G
APOSPC501980-1GEA
6850
CZK
APOSPC501980-1G
APOSPC501980-5G
2-(Difluoromethoxy)-4-fluoroaniline
2-(Difluoromethoxy)-4-fluoroaniline
Vzorec:
C₇H₆F₃NO MW: 177,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04969924 CAS číslo: 832740-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...