Tisk…

2-Amino-4-bromo-6-fluorobenzoic acid ≥98%

Dodavatel: Apollo Scientific

PC501979-100MG PC501979-250MG PC501979-1G
APOSPC501979-100MGEA 4470 CZK
APOSPC501979-100MG APOSPC501979-250MG APOSPC501979-1G
2-Amino-4-bromo-6-fluorobenzoic acid ≥98%
2-Amino-4-bromo-6-fluorobenzoic acid
Vzorec: C₇H₅BrFNO₂
MW: 234,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26386859
CAS číslo: 1312454-86-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR