Tisk…

6-Bromo-2,3-difluorobenzyl bromide 97%

Dodavatel: Apollo Scientific

PC501975-10G PC501975-1G PC501975-5G
APOSPC501975-10GEA 35380 CZK
APOSPC501975-10G APOSPC501975-1G APOSPC501975-5G
6-Bromo-2,3-difluorobenzyl bromide 97%
6-Bromo-2,3-difluorobenzyl bromide
Vzorec: C₇H₄Br₂F₂
MW: 285.911 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28060860
CAS číslo: 1807193-48-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR